2-Fluoro-3-isopropoxypyridine-5-boronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

24729-5G 24729-1G 24729-25G 24729-500MG
77632-226EA 942 USD
77632-226 77637-440 77654-410 77649-494
2-Fluoro-3-isopropoxypyridine-5-boronic acid pinacol ester ≥95%
2-Fluoro-3-isopropoxypyridine-5-boronic acid pinacol ester

Precaución: For Research Use Only

Formula: C₁₄H₂₁BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD26045245
CAS Number: 2121513-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR