2-Chloro-4-(1H-pyrazol-1-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

247257-5G 247257-25G 247257-1G 247257-500MG
77632-224EA 643 USD
77632-224 77658-200 77641-470 77649-492
2-Chloro-4-(1H-pyrazol-1-yl)benzaldehyde ≥97%
2-Chloro-4-(1H-pyrazol-1-yl)benzaldehyde

Precaución: For Research Use Only

Formula: C₁₀H₇ClN₂O
MW: 206.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11656800
CAS Number: 1186663-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR