1-Iodo-2-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

247241-5G 247241-1G 247241-500MG
77628-216EA 1546.03 USD
77628-216 77637-438 77649-490
1-Iodo-2-(methoxymethyl)benzene ≥97%
1-Iodo-2-(methoxymethyl)benzene

Precaución: For Research Use Only

Formula: 248.06 g/mol
MW: C₈H₉IO
Storage Temperature: Ambient
CAS Number: 90003-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR