1-Methoxy-3-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

247224-1G 247224-5G 247224-500MG
77634-478EA 1214 USD
77634-478 77658-198 77645-426
1-Methoxy-3-(methoxymethyl)benzene ≥97%
1-Methoxy-3-(methoxymethyl)benzene

Precaución: For Research Use Only

Formula: C₉H₁₂O₂
Storage Temperature: Ambient
CAS Number: 1515-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR