1-(Methoxymethyl)-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

247219-5G 247219-25G 247219-1G
77634-476EA 187 USD
77634-476 77658-196 77641-468
1-(Methoxymethyl)-4-(trifluoromethyl)benzene ≥97%
1-(Methoxymethyl)-4-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: C₉H₉F₃O
MW: 190.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11504959
CAS Number: 155820-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR