(2-Fluorobiphenyl-5-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

24711-5G 24711-1G 24711-25G 24711-500MG
77632-222EA 749 USD
77632-222 77637-436 77654-408 77649-488
(2-Fluorobiphenyl-5-yl)methanol ≥97%
(2-Fluorobiphenyl-5-yl)methanol

Precaución: For Research Use Only

Formula: 202.224 g/mol
MW: C₁₃H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD14701917
CAS Number: 89951-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR