3-Ethoxy-5-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

24670-5G 24670-25G 24670-1G 24670-500MG
77632-220EA 1198 USD
77632-220 77654-402 77641-466 77645-422
3-Ethoxy-5-methylpyridine ≥97%
3-Ethoxy-5-methylpyridine

Precaución: For Research Use Only

Formula: 137.179 g/mol
MW: C₈H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD18254212
CAS Number: 1256825-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR