1-(4-Fluoro-2-methoxyphenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

246642-5G 246642-1G 246642-500MG
77654-400EA 2250 USD
77654-400 77641-464 77649-484
1-(4-Fluoro-2-methoxyphenyl)-1-propanone ≥95%
1-(4-Fluoro-2-methoxyphenyl)-1-propanone

Precaución: For Research Use Only

Formula: 182.19 g/mol
MW: C₁₀H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD11617664
CAS Number: 231958-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR