Cyclohexyl-2-methoxyphenyl ketone ≥95%

Supplier: AOBChem USA

Warning

246614-5G 246614-1G 246614-25G 246614-500MG
77632-216EA 1412 USD
77632-216 77637-430 77654-398 77645-420
Cyclohexyl-2-methoxyphenyl ketone ≥95%
Cyclohexyl-2-methoxyphenyl ketone

Precaución: For Research Use Only

Formula: C₁₄H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD03841339
CAS Number: 111504-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR