Cyclobutyl(2,4-dimethylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

246520-5G 246520-25G 246520-1G 246520-500MG
77632-212EA 1412 USD
77632-212 77654-394 77641-458 77645-416
Cyclobutyl(2,4-dimethylphenyl)methanone ≥95%
Cyclobutyl(2,4-dimethylphenyl)methanone

Precaución: For Research Use Only

Formula: C₁₃H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD03841265
CAS Number: 898790-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR