6-Bromo-1-chloro-2-isopropoxynaphthalene ≥97%

Supplier: AOBChem USA

Warning

24651-5G 24651-1G 24651-500MG
77628-212EA 1484.23 USD
77628-212 77641-456 77649-480
6-Bromo-1-chloro-2-isopropoxynaphthalene ≥97%
6-Bromo-1-chloro-2-isopropoxynaphthalene

Precaución: For Research Use Only

Formula: 299.591 g/mol
MW: C₁₃H₁₂BrClO
Storage Temperature: Ambient
MDL Number: MFCD22543739
CAS Number: 1394291-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR