6-Bromo-1-chloro-2-ethoxynaphthalene ≥97%

Supplier: AOBChem USA

Warning

24650-5G 24650-1G 24650-25G 24650-500MG
77632-210EA 1380 USD
77632-210 77637-428 77654-392 77649-478
6-Bromo-1-chloro-2-ethoxynaphthalene ≥97%
6-Bromo-1-chloro-2-ethoxynaphthalene

Precaución: For Research Use Only

Formula: C₁₂H₁₀BrClO
Storage Temperature: Ambient
MDL Number: MFCD22543738
CAS Number: 1394291-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR