1-(4-Methoxy-2-methylphenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

246470-5G 246470-1G 246470-25G 246470-500MG
77632-206EA 767 USD
77632-206 77637-426 77658-190 77649-474
1-(4-Methoxy-2-methylphenyl)-1-propanone ≥95%
1-(4-Methoxy-2-methylphenyl)-1-propanone

Precaución: For Research Use Only

Formula: 178.23 g/mol
MW: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD11617652
CAS Number: 53773-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR