2-Fluorophenyl cyclopropyl ketone ≥97%

Supplier: AOBChem USA

Warning

246469-5G 246469-25G 246469-1G 246469-500MG
77632-204EA 1111 USD
77632-204 77658-188 77641-452 77645-414
2-Fluorophenyl cyclopropyl ketone ≥97%
2-Fluorophenyl cyclopropyl ketone

Precaución: For Research Use Only

Formula: C₁₀H₉FO
Storage Temperature: Ambient
MDL Number: MFCD03841228
CAS Number: 141030-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR