2',4'-Dimethoxypropiophenone ≥95%

Supplier: AOBChem USA

Warning

246459-5G 246459-25G
77634-472EA 86 USD
77634-472 77658-186
2',4'-Dimethoxypropiophenone ≥95%
2',4'-Dimethoxypropiophenone

Precaución: For Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00027012
CAS Number: 831-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR