1-(3-Fluoro-2-methylphenyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

246458-5G 246458-25G 246458-1G 246458-500MG
77632-202EA 1231 USD
77632-202 77658-184 77641-450 77649-472
1-(3-Fluoro-2-methylphenyl)-1-propanone ≥95%
1-(3-Fluoro-2-methylphenyl)-1-propanone

Precaución: For Research Use Only

Formula: 166.19 g/mol
MW: C₁₀H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD11617665
CAS Number: 1261852-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR