1-(2-Fluorophenyl)-2-methylpropan-1-one ≥95%

Supplier: AOBChem USA

Warning

246447-5G 246447-25G 246447-1G 246447-500MG
77632-200EA 768 USD
77632-200 77654-388 77641-448 77649-470
1-(2-Fluorophenyl)-2-methylpropan-1-one ≥95%
1-(2-Fluorophenyl)-2-methylpropan-1-one

Precaución: For Research Use Only

Formula: C₁₀H₁₁FO
MW: 166.1922 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16301198
CAS Number: 116707-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR