1-(2-Methoxyphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

246427-5G 246427-25G
77634-470EA 64 USD
77634-470 77654-386
1-(2-Methoxyphenyl)propan-1-one ≥95%
1-(2-Methoxyphenyl)propan-1-one

Precaución: For Research Use Only

Formula: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00048613
CAS Number: 5561-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR