α-Cyclopropyl-2,5-difluoro-α-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

246242-5G 246242-25G 246242-1G 246242-500MG
77628-208EA 1484.23 USD
77628-208 77635-554 77641-446 77645-412
α-Cyclopropyl-2,5-difluoro-α-methylbenzenemethanol ≥95%
α-Cyclopropyl-2,5-difluoro-α-methylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₁H₁₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD17223840
CAS Number: 1342142-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR