α-Cyclopropyl-2,6-difluoro-α-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

246178-5G 246178-25G 246178-1G 246178-500MG
77628-206EA 1484.23 USD
77628-206 77635-552 77641-444 77645-410
α-Cyclopropyl-2,6-difluoro-α-methylbenzenemethanol ≥95%
α-Cyclopropyl-2,6-difluoro-α-methylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₁H₁₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD19637743
CAS Number: 1343591-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR