α-Cyclopropyl-α,4-dimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

246096-5G 246096-25G 246096-1G
77634-468EA 214 USD
77634-468 77654-384 77641-442
α-Cyclopropyl-α,4-dimethylbenzenemethanol ≥95%
α-Cyclopropyl-α,4-dimethylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₂H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD00065230
CAS Number: 33446-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR