4-Methyl-6-methylsulfanyl-nicotinic acid ≥97%

Supplier: AOBChem USA

Warning

24584-5G 24584-1G 24584-25G 24584-500MG
77632-198EA 1380 USD
77632-198 77637-420 77654-380 77645-408
4-Methyl-6-methylsulfanyl-nicotinic acid ≥97%
4-Methyl-6-methylsulfanyl-nicotinic acid

Precaución: For Research Use Only

Formula: 183.228 g/mol
MW: C₈H₉NO₂S
Storage Temperature: Ambient
MDL Number: MFCD21088019
CAS Number: 1355177-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR