3-Bromo-5-(methylsulfanyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

24578-5G 24578-25G 24578-1G 24578-500MG
77632-196EA 656 USD
77632-196 77654-378 77641-440 77649-464
3-Bromo-5-(methylsulfanyl)phenylboronic acid ≥97%
3-Bromo-5-(methylsulfanyl)phenylboronic acid

Precaución: For Research Use Only

Formula: 246.917 g/mol
MW: C₇H₈BBrO₂S
Storage Temperature: Ambient
MDL Number: MFCD22628200
CAS Number: 1451392-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR