3-Chloro-5-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

24565-5G 24565-25G 24565-1G 24565-500MG
77632-194EA 841 USD
77632-194 77658-182 77641-438 77645-406
3-Chloro-5-(methylthio)benzaldehyde ≥97%
3-Chloro-5-(methylthio)benzaldehyde

Precaución: For Research Use Only

Formula: 186.659 g/mol
MW: C₈H₇ClOS
Storage Temperature: Ambient
MDL Number: MFCD28650660
CAS Number: 1289164-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR