2,5-Difluoro-3-methylphenol ≥97%

Supplier: AOBChem USA

Warning

245328-5G 245328-1G 245328-500MG
77628-204EA 1484.23 USD
77628-204 77641-436 77645-404
2,5-Difluoro-3-methylphenol ≥97%
2,5-Difluoro-3-methylphenol

Precaución: For Research Use Only

Formula: C₇H₆F₂O
Storage Temperature: Ambient
MDL Number: MFCD28790487
CAS Number: 1803786-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR