(3-Chloro-5-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

24529-5G 24529-25G 24529-1G 24529-500MG
77632-192EA 803 USD
77632-192 77658-180 77641-434 77645-402
(3-Chloro-5-methylphenyl)methanol ≥97%
(3-Chloro-5-methylphenyl)methanol

Precaución: For Research Use Only

Formula: 156.609 g/mol
MW: C₈H₉ClO
Storage Temperature: Ambient
MDL Number: MFCD18393675
CAS Number: 116069-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR