1-Bromo-2-isopropoxy-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

24526-5G 24526-25G 24526-1G 24526-500MG
77632-190EA 760 USD
77632-190 77654-376 77641-432 77649-462
1-Bromo-2-isopropoxy-4-methylbenzene ≥97%
1-Bromo-2-isopropoxy-4-methylbenzene

Precaución: For Research Use Only

Formula: 229.114 g/mol
MW: C₁₀H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD22543719
CAS Number: 221111-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR