1-Methoxy-2,4-bis(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

24410-5G 24410-25G 24410-1G 24410-500MG
77632-188EA 841 USD
77632-188 77654-372 77641-428 77649-458
1-Methoxy-2,4-bis(trifluoromethyl)benzene ≥97%
1-Methoxy-2,4-bis(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: 244.134 g/mol
MW: C₉H₆F₆O
Storage Temperature: Ambient
MDL Number: MFCD13185339
CAS Number: 577-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR