1-Bromo-2,4-diethylbenzene ≥97%

Supplier: AOBChem USA

Warning

243660-1G 243660-5G 243660-500MG
77637-414EA 574 USD
77637-414 77654-368 77645-398
1-Bromo-2,4-diethylbenzene ≥97%
1-Bromo-2,4-diethylbenzene

Precaución: For Research Use Only

Formula: 213.11 g/mol
MW: C₁₀H₁₃Br
Storage Temperature: Ambient
CAS Number: 90267-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR