2,5-Diethylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

243648-5G 243648-1G 243648-25G 243648-500MG
77632-184EA 812 USD
77632-184 77637-412 77658-174 77649-452
2,5-Diethylbenzoic acid ≥95%
2,5-Diethylbenzoic acid

Precaución: For Research Use Only

Formula: 178.23 g/mol
MW: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD11557890
CAS Number: 474022-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR