3-Chloro-4-methylsulfanyl-benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

24358-5G 24358-1G 24358-25G 24358-500MG
77632-182EA 651 USD
77632-182 77637-410 77658-172 77645-396
3-Chloro-4-methylsulfanyl-benzaldehyde ≥97%
3-Chloro-4-methylsulfanyl-benzaldehyde

Precaución: For Research Use Only

Formula: 186.659 g/mol
MW: C₈H₇ClOS
Storage Temperature: Ambient
MDL Number: MFCD24056861
CAS Number: 38125-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR