1-(4-Chloro-3-ethoxy-2-fluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

243437-5G 243437-1G 243437-25G 243437-500MG
77628-200EA 1484.23 USD
77628-200 77637-408 77658-170 77649-450
1-(4-Chloro-3-ethoxy-2-fluorophenyl)ethanone ≥95%
1-(4-Chloro-3-ethoxy-2-fluorophenyl)ethanone

Precaución: For Research Use Only

Formula: 216.637 g/mol
MW: C₁₀H₁₀ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD30717176
CAS Number: 2169573-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR