3-Bromo-1-tosyl-1H-pyrrolo[2,3-b]pyridine ≥95%

Supplier: AOBChem USA

Warning

24280-5G 24280-25G
77634-460EA 59 USD
77634-460 77658-164
3-Bromo-1-tosyl-1H-pyrrolo[2,3-b]pyridine ≥95%
3-Bromo-1-tosyl-1H-pyrrolo[2,3-b]pyridine

Precaución: For Research Use Only

Formula: C₁₄H₁₁BrN₂O₂S
MW: 351.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10697518
CAS Number: 226085-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR