α-Heptyl-2-methylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

242308-5G 242308-1G 242308-25G 242308-500MG
77632-178EA 1381 USD
77632-178 77637-406 77654-366 77649-444
α-Heptyl-2-methylbenzenemethanol ≥95%
α-Heptyl-2-methylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₅H₂₄O
Storage Temperature: Ambient
MDL Number: MFCD18352315
CAS Number: 1340566-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR