Methyl-5-chloro-6-fluoronicotinate ≥95%

Supplier: AOBChem USA

Warning

24183-5G 24183-25G 24183-1G 24183-500MG
77632-176EA 770 USD
77632-176 77658-162 77645-394 77649-442
Methyl-5-chloro-6-fluoronicotinate ≥95%
Methyl-5-chloro-6-fluoronicotinate

Precaución: For Research Use Only

Formula: C₇H₅ClFNO₂
Storage Temperature: Ambient
MDL Number: MFCD12498696
CAS Number: 1171918-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR