3-Chloro-2-isopropoxypyridine-5-boronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

24159-1G 24159-5G 24159-500MG
77637-402EA 652 USD
77637-402 77654-364 77649-440
3-Chloro-2-isopropoxypyridine-5-boronic acid pinacol ester ≥95%
3-Chloro-2-isopropoxypyridine-5-boronic acid pinacol ester

Precaución: For Research Use Only

Formula: 297.585 g/mol
MW: C₁₄H₂₁BClNO₃
Storage Temperature: Ambient
MDL Number: MFCD16996490
CAS Number: 1344997-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR