4-Chloro-2-fluoro-6-methylaniline ≥97%

Supplier: AOBChem USA

Warning

241481-5G 241481-1G 241481-25G 241481-500MG
77632-174EA 698 USD
77632-174 77637-400 77654-362 77649-438
4-Chloro-2-fluoro-6-methylaniline ≥97%
4-Chloro-2-fluoro-6-methylaniline

Precaución: For Research Use Only

Formula: C₇H₇ClFN
MW: 159.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16817663
CAS Number: 939989-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR