1-(3-Methylthiophen-2-yl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

241476-5G 241476-1G 241476-500MG
77654-360EA 2001 USD
77654-360 77641-422 77645-390
1-(3-Methylthiophen-2-yl)propan-1-one ≥95%
1-(3-Methylthiophen-2-yl)propan-1-one

Precaución: For Research Use Only

Formula: 154.2294 g/mol
MW: C₈H₁₀OS
Storage Temperature: Ambient
MDL Number: MFCD11870141
CAS Number: 59303-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR