1-(4-Bromo-3-chlorophenyl)-2-methylpropan-1-one ≥95%

Supplier: AOBChem USA

Warning

241472-5G 241472-1G 241472-500MG
77654-358EA 2001 USD
77654-358 77641-420 77649-436
1-(4-Bromo-3-chlorophenyl)-2-methylpropan-1-one ≥95%
1-(4-Bromo-3-chlorophenyl)-2-methylpropan-1-one

Precaución: For Research Use Only

Formula: C₁₀H₁₀BrClO
Storage Temperature: Ambient
MDL Number: MFCD28107186
CAS Number: 1592443-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR