1-Fluoro-2-methyl-3-(methylsulfanyl)benzene ≥95%

Supplier: AOBChem USA

Warning

24106-5G 24106-1G 24106-500MG
77654-356EA 1961 USD
77654-356 77641-416 77645-388
1-Fluoro-2-methyl-3-(methylsulfanyl)benzene ≥95%
1-Fluoro-2-methyl-3-(methylsulfanyl)benzene

Precaución: For Research Use Only

Formula: 156.22 g/mol
MW: C₈H₉FS
Storage Temperature: Ambient
MDL Number: MFCD11617786
CAS Number: 134646-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR