6-Acetamido-4-methylpyridine-3-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

24060-1G 24060-5G 24060-500MG
77637-398EA 694 USD
77637-398 77658-158 77649-432
6-Acetamido-4-methylpyridine-3-boronic acid pinacol ester ≥97%
6-Acetamido-4-methylpyridine-3-boronic acid pinacol ester

Precaución: For Research Use Only

Formula: 276.139 g/mol
MW: C₁₄H₂₁BN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD18206563
CAS Number: 1351995-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR