2-Fluoro-5-(methylsulfonyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

24047-1G 24047-5G 24047-500MG
77637-396EA 599 USD
77637-396 77654-354 77649-430
2-Fluoro-5-(methylsulfonyl)pyridine ≥97%
2-Fluoro-5-(methylsulfonyl)pyridine

Precaución: For Research Use Only

Formula: 175.181 g/mol
MW: C₆H₆FNO₂S
Storage Temperature: Ambient
MDL Number: MFCD19689810
CAS Number: 1037764-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR