6-[4-(1,1-Dimethylethyl)phenyl]-3-pyridinamine ≥97%

Supplier: AOBChem USA

Warning

240029-1G 240029-5G 240029-500MG
77634-454EA 882 USD
77634-454 77658-154 77645-386
6-[4-(1,1-Dimethylethyl)phenyl]-3-pyridinamine ≥97%
6-[4-(1,1-Dimethylethyl)phenyl]-3-pyridinamine

Precaución: For Research Use Only

Formula: C₁₅H₁₈N₂
Storage Temperature: Ambient
MDL Number: MFCD16314017
CAS Number: 1110656-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR