3-(2-Aminopyrimidin-5-yl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

238718-5G 238718-1G 238718-25G 238718-500MG
77632-168EA 971 USD
77632-168 77637-392 77658-150 77649-420
3-(2-Aminopyrimidin-5-yl)benzoic acid ≥97%
3-(2-Aminopyrimidin-5-yl)benzoic acid

Precaución: For Research Use Only

Formula: C₁₁H₉N₃O₂
MW: 215.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05864813
CAS Number: 914349-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR