3-Ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

238436-5G 238436-25G 238436-1G 238436-500MG
77632-164EA 1284 USD
77632-164 77658-148 77641-406 77645-382
3-Ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole ≥97%
3-Ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

Precaución: For Research Use Only

Formula: 272.15 g/mol
MW: C₁₅H₂₁BN₂O₂
Storage Temperature: Ambient
CAS Number: 864774-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR