2,6-Dimethyl-2',4'-difluorobiphenyl-4-carboxaldehyde ≥95%

Supplier: AOBChem USA

Warning

23836-1G 23836-500MG 23836-5G
77637-390EA 565 USD
77637-390 77652-532 77658-146
2,6-Dimethyl-2',4'-difluorobiphenyl-4-carboxaldehyde ≥95%
2,6-Dimethyl-2',4'-difluorobiphenyl-4-carboxaldehyde

Precaución: For Research Use Only

Formula: C₁₅H₁₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD22543674
CAS Number: 1350760-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR