2,6-Dimethyl-4'-fluorobiphenyl-4-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

23834-1G 23834-5G 23834-500MG
77637-388EA 565 USD
77637-388 77654-346 77649-416
2,6-Dimethyl-4'-fluorobiphenyl-4-carboxaldehyde ≥97%
2,6-Dimethyl-4'-fluorobiphenyl-4-carboxaldehyde

Precaución: For Research Use Only

Formula: 228.262 g/mol
MW: C₁₅H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD22543673
CAS Number: 1350760-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR