4-(Dimethylaminomethyl)-6-fluoro-2-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

23800-5G 23800-25G 23800-1G
77634-450EA 184 USD
77634-450 77654-344 77641-404
4-(Dimethylaminomethyl)-6-fluoro-2-methoxyphenol ≥97%
4-(Dimethylaminomethyl)-6-fluoro-2-methoxyphenol

Precaución: For Research Use Only

Formula: C₁₀H₁₄FNO₂
Storage Temperature: Ambient
MDL Number: MFCD04972107
CAS Number: 103905-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR