6-(3-Trifluoromethylphenyl)pyridine-3-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

23705-5G 23705-1G 23705-500MG
77654-342EA 1836 USD
77654-342 77641-402 77645-380
6-(3-Trifluoromethylphenyl)pyridine-3-carboxaldehyde ≥97%
6-(3-Trifluoromethylphenyl)pyridine-3-carboxaldehyde

Precaución: For Research Use Only

Formula: 251.204 g/mol
MW: C₁₃H₈F₃NO
Storage Temperature: Ambient
MDL Number: MFCD04115429
CAS Number: 898405-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR