3-Bromo-5-(3-fluorophenyl)-pyridine ≥95%

Supplier: AOBChem USA

Warning

23670-5G 23670-500MG 23670-25G 23670-1G
77632-162EA 692 USD
77632-162 77652-530 77654-340 77641-400
3-Bromo-5-(3-fluorophenyl)-pyridine ≥95%
3-Bromo-5-(3-fluorophenyl)-pyridine

Precaución: For Research Use Only

Formula: 252.082 g/mol
MW: C₁₁H₇BrFN
Storage Temperature: Ambient
MDL Number: MFCD19544000
CAS Number: 1276123-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR